siRNA Details
| Gene Symbol: Zfp622 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009652.1 | NP_001009652.1 | 1319 | |||
| GenBank mRNA | BC088214.1 | - | 1319 | 7383802 | ||
| Gene Symbol: LOC103690028 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760822.1 | XP_008759044.1 | 5 | 2457 | ||

Zfp622
Genomic Map