siRNA Details
| Gene Symbol: Dnah5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760824.1 | XP_008759046.1 | 4677 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dnah5| Gene Symbol: Dnah5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760824.1 | XP_008759046.1 | 4677 | |||