siRNA Details
| Gene Symbol: March6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760826.1 | XP_008759048.1 | 5, 6 | 675 | ||
| XM_215517.8 | XP_215517.5 | 5, 6 | 620 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

March6| Gene Symbol: March6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760826.1 | XP_008759048.1 | 5, 6 | 675 | ||
| XM_215517.8 | XP_215517.5 | 5, 6 | 620 | |||