siRNA Details
| Gene Symbol: Ralyl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024978.1 | NP_001020149.1 | 5 | 443 | ||
| XM_006232123.2 | XP_006232185.1 | 6 | 768 | |||
| XM_006232126.2 | XP_006232188.1 | 6 | 1265 | |||
| XM_008760852.1 | XP_008759074.1 | 6 | 768 | |||
| XM_008760853.1 | XP_008759075.1 | 6 | 1265 | |||
| GenBank mRNA | BC092614.1 | - | 5 | 443 | 7311942 | |

Ralyl
Genomic Map