siRNA Details
| Gene Symbol: tGap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007635.2 | NP_001007636.2 | 26 | 3733 | ||
| XM_006232134.2 | XP_006232196.1 | 28 | 4075 | |||
| GenBank mRNA | AY631396.1 | - | 26 | 3733 | ||
| AY631398.1 | - | 18 | 2672 | |||
| AY631399.1 | - | 26 | 3498 | |||
| AY631400.1 | - | 22 | 2872 | |||

tGap1
Genomic Map