siRNA Details
| Gene Symbol: Tpd52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006232140.1 | XP_006232202.1 | 4, 5 | 801 | ||
| XM_006232143.2 | XP_006232205.1 | 4, 5 | 1190 | |||
| XM_008760861.1 | XP_008759083.1 | 4, 5 | 801 | |||
| XM_008760863.1 | XP_008759085.1 | 4, 5 | 1190 | |||

Tpd52
Genomic Map