siRNA Details
| Gene Symbol: WNT8B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003393.3 | NP_003384.2 | 4 | 412 | ||
| GenBank mRNA | AB073637.1 | - | 4 | 410 | ||
| CN276452.1 | - | 4 | 413 | |||
| X91940.1 | - | 3 | 232 | |||
| Y11094.1 | - | 4 | 435 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

WNT8B| Gene Symbol: WNT8B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003393.3 | NP_003384.2 | 4 | 412 | ||
| GenBank mRNA | AB073637.1 | - | 4 | 410 | ||
| CN276452.1 | - | 4 | 413 | |||
| X91940.1 | - | 3 | 232 | |||
| Y11094.1 | - | 4 | 435 | |||