siRNA Details
| Gene Symbol: Gnb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013910.2 | NP_001013932.1 | 7 | 1031 | ||
| GenBank mRNA | AY552804.1 | - | 7 | 958 | ||
| BC097370.1 | - | 7 | 1031 | 7444022 | ||
| DQ120489.1 | - | 7 | 923 | |||
| DQ120490.1 | - | 7 | 923 | |||

Gnb4
Genomic Map