siRNA Details
| Gene Symbol: Hspa4l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106428.2 | NP_001099898.2 | 1 | 119 | ||
| XM_006232297.2 | XP_006232359.1 | 1 | 206 | |||
| GenBank mRNA | BC168157.1 | - | 1 | 119 | 8363916 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hspa4l| Gene Symbol: Hspa4l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106428.2 | NP_001099898.2 | 1 | 119 | ||
| XM_006232297.2 | XP_006232359.1 | 1 | 206 | |||
| GenBank mRNA | BC168157.1 | - | 1 | 119 | 8363916 | |