siRNA Details
| Gene Symbol: RGD1307595 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134511.1 | NP_001127983.1 | 1 | 133 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1307595| Gene Symbol: RGD1307595 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134511.1 | NP_001127983.1 | 1 | 133 | ||