siRNA Details
| Gene Symbol: Rexo1l1p | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006232358.2 | XP_006232420.1 | 1 | 888 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rexo1l1p| Gene Symbol: Rexo1l1p | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006232358.2 | XP_006232420.1 | 1 | 888 | ||