siRNA Details
| Gene Symbol: Spg20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106433.2 | NP_001099903.2 | 2 | 216 | ||
| XM_006232365.2 | XP_006232427.1 | 2 | 544 | |||
| XM_006232366.2 | XP_006232428.1 | 2 | 535 | |||
| XM_006232367.2 | XP_006232429.1 | 2 | 333 | |||

Spg20
Genomic Map