siRNA Details
| Gene Symbol: Tm4sf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106434.1 | NP_001099904.1 | 3 | 693 | ||
| XM_008760989.1 | XP_008759211.1 | 3 | 820 | |||
| GenBank mRNA | FQ220762.1 | - | 3 | 459 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tm4sf1| Gene Symbol: Tm4sf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106434.1 | NP_001099904.1 | 3 | 693 | ||
| XM_008760989.1 | XP_008759211.1 | 3 | 820 | |||
| GenBank mRNA | FQ220762.1 | - | 3 | 459 | ||