siRNA Details
| Gene Symbol: RGD1560324 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106436.1 | NP_001099906.1 | 4 | 421 | ||
| XM_008760990.1 | XP_008759212.1 | 3 | 421 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1560324| Gene Symbol: RGD1560324 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106436.1 | NP_001099906.1 | 4 | 421 | ||
| XM_008760990.1 | XP_008759212.1 | 3 | 421 | |||