siRNA Details
| Gene Symbol: LOC100910567 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749284.2 | XP_003749332.1 | 2 | 394 | ||
| Gene Symbol: Aadacl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191706.1 | NP_001178635.1 | 282 | |||

LOC100910567
Genomic Map