siRNA Details
| Gene Symbol: Gmps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024754.1 | NP_001019925.1 | 9 | 1178 | ||
| GenBank mRNA | BC098016.1 | - | 9 | 1178 | 7454908 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gmps| Gene Symbol: Gmps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024754.1 | NP_001019925.1 | 9 | 1178 | ||
| GenBank mRNA | BC098016.1 | - | 9 | 1178 | 7454908 | |