siRNA Details
| Gene Symbol: Vom2r45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008761059.1 | XP_008759281.1 | 3 | 3564 | ||
| XM_215580.6 | XP_215580.2 | 3 | 3546 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vom2r45| Gene Symbol: Vom2r45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008761059.1 | XP_008759281.1 | 3 | 3564 | ||
| XM_215580.6 | XP_215580.2 | 3 | 3546 | |||
Set Membership: |
|