siRNA Details
| Gene Symbol: Schip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100666.1 | NP_001094136.1 | 6, 7 | 1162 | ||
| XM_008761079.1 | XP_008759301.1 | 3, 4 | 4100 | |||
| XM_008761080.1 | XP_008759302.1 | 3, 4 | 969 | |||
| XM_008761081.1 | XP_008759303.1 | 3, 4 | 845 | |||
| XM_008761082.1 | XP_008759304.1 | 2, 3 | 3117 | |||
| XM_008761083.1 | XP_008759305.1 | 2, 3 | 416 | |||
| GenBank mRNA | BC092618.1 | - | 2, 3 | 134 | ||
| BC169016.1 | - | 2, 3 | 363 | |||

Schip1
Genomic Map