siRNA Details
Gene Symbol: Schip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100666.1 | NP_001094136.1 | 9 | 1512 | ||
XM_008761079.1 | XP_008759301.1 | 6 | 4450 | |||
XM_008761080.1 | XP_008759302.1 | 6 | 1319 | |||
XM_008761081.1 | XP_008759303.1 | 6 | 1195 | |||
XM_008761082.1 | XP_008759304.1 | 5 | 3467 | |||
XM_008761083.1 | XP_008759305.1 | 5 | 766 | |||
GenBank mRNA | BC092618.1 | - | 5 | 484 | ||
BC169016.1 | - | 5 | 713 |
