siRNA Details
| Gene Symbol: WNT9A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003395.2 | NP_003386.1 | 4 | 828 | ||
| XM_011544271.2 | XP_011542573.1 | 4 | 1311 | |||
| GenBank mRNA | AB060283.1 | - | 4 | 751 | ||
| BC111960.1 | - | 4 | 828 | 8327428 | ||
| BC113431.1 | - | 4 | 828 | 8322483 | ||

WNT9A
Genomic Map