siRNA Details
| Gene Symbol: Ift80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013911.1 | NP_001013933.1 | 3 | 323 | ||
| GenBank mRNA | BC081931.1 | - | 3 | 323 | 7113994 | |
| BC081937.1 | - | 3 | 323 | 7115382 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ift80| Gene Symbol: Ift80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013911.1 | NP_001013933.1 | 3 | 323 | ||
| GenBank mRNA | BC081931.1 | - | 3 | 323 | 7113994 | |
| BC081937.1 | - | 3 | 323 | 7115382 | ||