siRNA Details
| Gene Symbol: LOC102550932 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006232552.2 | XP_006232614.1 | 1 | 1728 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102550932| Gene Symbol: LOC102550932 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006232552.2 | XP_006232614.1 | 1 | 1728 | ||