siRNA Details
| Gene Symbol: Glmp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004226.1 | NP_001004226.1 | 2 | 367 | ||
| XM_008761133.1 | XP_008759355.1 | 2 | 488 | |||
| GenBank mRNA | BC079294.1 | - | 2 | 367 | 7134483 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Glmp| Gene Symbol: Glmp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004226.1 | NP_001004226.1 | 2 | 367 | ||
| XM_008761133.1 | XP_008759355.1 | 2 | 488 | |||
| GenBank mRNA | BC079294.1 | - | 2 | 367 | 7134483 | |