siRNA Details
| Gene Symbol: Dap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011950.2 | NP_001011950.2 | 12 | 1349 | ||
| XM_006232610.2 | XP_006232672.1 | 11 | 1055 | |||
| XM_006232611.2 | XP_006232673.1 | 12 | 1153 | |||
| GenBank mRNA | BC085878.1 | - | 12 | 1349 | 7191747 | |
| FQ230674.1 | - | 12 | 1138 | |||

Dap3
Genomic Map