siRNA Details
| Gene Symbol: Dcst1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008761243.1 | XP_008759465.1 | 10 | 1611 | ||
| GenBank mRNA | FQ210715.1 | - | 1771 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dcst1| Gene Symbol: Dcst1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008761243.1 | XP_008759465.1 | 10 | 1611 | ||
| GenBank mRNA | FQ210715.1 | - | 1771 | |||