siRNA Details
| Gene Symbol: Dcst2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008761239.1 | XP_008759461.1 | 8 | 1252 | ||
| XM_008761240.1 | XP_008759462.1 | 8 | 1252 | |||
| XM_008761241.1 | XP_008759463.1 | 8 | 1177 | |||
| XM_008761242.1 | XP_008759464.1 | 8 | 1144 | |||

Dcst2
Genomic Map