siRNA Details
Gene Symbol: Mllt11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013912.1 | NP_001013934.1 | 2 | 812 | ||
GenBank mRNA | BC087583.1 | - | 2 | 812 | 7316660 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mllt11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013912.1 | NP_001013934.1 | 2 | 812 | ||
GenBank mRNA | BC087583.1 | - | 2 | 812 | 7316660 |