siRNA Details
| Gene Symbol: Sf3b4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011951.1 | NP_001011951.1 | 3 | 775 | ||
| GenBank mRNA | BC078997.1 | - | 3 | 775 | 7112738 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sf3b4| Gene Symbol: Sf3b4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011951.1 | NP_001011951.1 | 3 | 775 | ||
| GenBank mRNA | BC078997.1 | - | 3 | 775 | 7112738 | |