siRNA Details
| Gene Symbol: Rbm8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271138.1 | NP_001258067.1 | 2, 3 | 167 | ||
| GenBank mRNA | DQ359102.1 | - | 2, 3 | 119 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rbm8a| Gene Symbol: Rbm8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271138.1 | NP_001258067.1 | 2, 3 | 167 | ||
| GenBank mRNA | DQ359102.1 | - | 2, 3 | 119 | ||