siRNA Details
| Gene Symbol: Acp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031645.1 | NP_001026815.1 | 4 | 478 | ||
| GenBank mRNA | BC099131.1 | - | 4 | 478 | 7444695 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Acp6| Gene Symbol: Acp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031645.1 | NP_001026815.1 | 4 | 478 | ||
| GenBank mRNA | BC099131.1 | - | 4 | 478 | 7444695 | |