siRNA Details
| Gene Symbol: Tbx15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106451.1 | NP_001099921.1 | 3 | 473 | ||
| XM_006233027.2 | XP_006233089.1 | 4 | 1048 | |||
| XM_008761285.1 | XP_008759507.1 | 4 | 5347 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tbx15| Gene Symbol: Tbx15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106451.1 | NP_001099921.1 | 3 | 473 | ||
| XM_006233027.2 | XP_006233089.1 | 4 | 1048 | |||
| XM_008761285.1 | XP_008759507.1 | 4 | 5347 | |||