siRNA Details
| Gene Symbol: Mab21l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106456.1 | NP_001099926.1 | 7 | 1423 | ||
| XM_006233045.2 | XP_006233107.1 | 7 | 1334 | |||
| XM_006233046.2 | XP_006233108.1 | 7 | 1618 | |||
| XM_008761374.1 | XP_008759596.1 | 7 | 1315 | |||
| XM_008761375.1 | XP_008759597.1 | 7 | 1239 | |||

Mab21l3
Genomic Map