siRNA Details
| Gene Symbol: Ppm1j | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005540.1 | NP_001005540.1 | 5 | 854 | ||
| XM_008761380.1 | XP_008759602.1 | 6 | 1352 | |||
| XM_008761381.1 | XP_008759603.1 | 6 | 1352 | |||
| GenBank mRNA | BC082053.1 | - | 5 | 854 | 7132147 | |

Ppm1j
Genomic Map