siRNA Details
| Gene Symbol: St7l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007639.1 | NP_001007640.1 | 12 | 1955 | ||
| GenBank mRNA | BC079194.1 | - | 12 | 1955 | 7131380 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

St7l| Gene Symbol: St7l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007639.1 | NP_001007640.1 | 12 | 1955 | ||
| GenBank mRNA | BC079194.1 | - | 12 | 1955 | 7131380 | |