siRNA Details
Gene Symbol: Slc16a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013913.1 | NP_001013935.1 | 5, 6 | 706 | ||
XM_008761384.1 | XP_008759606.1 | 5, 6 | 877 | |||
XM_008761385.1 | XP_008759607.1 | 5, 6 | 880 | |||
XM_008761386.1 | XP_008759608.1 | 5, 6 | 875 | |||
XM_008761387.1 | XP_008759609.1 | 5, 6 | 875 | |||
XM_008761390.1 | XP_008759612.1 | 5, 6 | 870 | |||
GenBank mRNA | BC085920.1 | - | 5, 6 | 706 | 7108742 |
