siRNA Details
| Gene Symbol: Lamtor5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106462.1 | NP_001099932.1 | 3, 4 | 432 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lamtor5| Gene Symbol: Lamtor5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106462.1 | NP_001099932.1 | 3, 4 | 432 | ||