siRNA Details
| Gene Symbol: Amigo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_206881.1 | NP_996764.1 | 1 | 869 | ||
| XM_006233140.2 | XP_006233202.1 | 2 | 1269 | |||
| XM_008761391.1 | XP_008759613.1 | 2 | 1304 | |||
| GenBank mRNA | AY237729.1 | - | 1 | 869 | ||
| BC167749.1 | - | 1 | 913 | 9021924 | ||

Amigo1
Genomic Map