siRNA Details
| Gene Symbol: Ntng1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106465.1 | NP_001099935.1 | 3 | 1671 | ||
| XM_008761393.1 | XP_008759615.1 | 1 | 837 | |||
| XM_008761394.1 | XP_008759616.1 | 1 | 837 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ntng1| Gene Symbol: Ntng1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106465.1 | NP_001099935.1 | 3 | 1671 | ||
| XM_008761393.1 | XP_008759615.1 | 1 | 837 | |||
| XM_008761394.1 | XP_008759616.1 | 1 | 837 | |||