siRNA Details
| Gene Symbol: XK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021083.2 | NP_066569.1 | 3 | 672 | ||
| GenBank mRNA | AY534238.1 | - | 3 | 796 | ||
| BC036019.1 | - | 3 | 713 | 5272840 | ||
| Z32684.2 | - | 3 | 672 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

XK| Gene Symbol: XK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021083.2 | NP_066569.1 | 3 | 672 | ||
| GenBank mRNA | AY534238.1 | - | 3 | 796 | ||
| BC036019.1 | - | 3 | 713 | 5272840 | ||
| Z32684.2 | - | 3 | 672 | |||