siRNA Details
| Gene Symbol: Dph5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017449.1 | NP_001017449.1 | 4 | 634 | ||
| XM_006233214.1 | XP_006233276.1 | 4 | 447 | |||
| XM_008761454.1 | XP_008759676.1 | 3 | 495 | |||
| XM_008761455.1 | XP_008759677.1 | 3 | 496 | |||
| GenBank mRNA | BC092598.1 | - | 4 | 634 | 7321341 | |

Dph5
Genomic Map