siRNA Details
| Gene Symbol: Hiat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106467.3 | NP_001099937.1 | 307 | |||
| GenBank mRNA | BC160912.1 | - | 296 | 7456962 | ||
| Gene Symbol: NEWGENE_1305281 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006233223.2 | XP_006233285.1 | 2, 3 | 439 | ||
| XM_006233224.2 | XP_006233286.1 | 2, 3 | 554 | |||
| XM_008761473.1 | XP_008759695.1 | 2, 3 | 310 | |||

Hiat1
Genomic Map