siRNA Details
Gene Symbol: Slc44a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013914.1 | NP_001013936.1 | 14 | 1898 | ||
GenBank mRNA | BC079142.1 | - | 14 | 1898 | 7128044 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc44a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013914.1 | NP_001013936.1 | 14 | 1898 | ||
GenBank mRNA | BC079142.1 | - | 14 | 1898 | 7128044 |