siRNA Details
Gene Symbol: Ccdc109b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006233280.2 | XP_006233342.1 | 995 | |||
XM_006233281.2 | XP_006233343.2 | 1489 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ccdc109b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006233280.2 | XP_006233342.1 | 995 | |||
XM_006233281.2 | XP_006233343.2 | 1489 |