siRNA Details
| Gene Symbol: Tnni3k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181769.1 | NP_861434.1 | 2 | 109 | ||
| XM_008761563.1 | XP_008759785.1 | 2 | 273 | |||
| GenBank mRNA | AY303692.1 | - | 2 | 109 | ||
| BC098648.1 | - | 2 | 114 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tnni3k| Gene Symbol: Tnni3k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181769.1 | NP_861434.1 | 2 | 109 | ||
| XM_008761563.1 | XP_008759785.1 | 2 | 273 | |||
| GenBank mRNA | AY303692.1 | - | 2 | 109 | ||
| BC098648.1 | - | 2 | 114 | |||