siRNA Details
| Gene Symbol: Depdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006233537.2 | XP_006233599.1 | 4 | 691 | ||
| XM_227798.8 | XP_227798.5 | 4 | 693 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Depdc1| Gene Symbol: Depdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006233537.2 | XP_006233599.1 | 4 | 691 | ||
| XM_227798.8 | XP_227798.5 | 4 | 693 | |||