siRNA Details
| Gene Symbol: Pigk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011953.1 | NP_001011953.1 | 5 | 417 | ||
| XM_008761549.1 | XP_008759771.1 | 5 | 443 | |||
| GenBank mRNA | BC083636.1 | - | 5 | 417 | 7134753 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pigk| Gene Symbol: Pigk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011953.1 | NP_001011953.1 | 5 | 417 | ||
| XM_008761549.1 | XP_008759771.1 | 5 | 443 | |||
| GenBank mRNA | BC083636.1 | - | 5 | 417 | 7134753 | |