siRNA Details
| Gene Symbol: Orc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199092.1 | NP_954523.1 | 9 | 835 | ||
| XM_006234150.2 | XP_006234212.1 | 11 | 999 | |||
| XM_006234151.2 | XP_006234213.1 | 10 | 925 | |||
| XM_006234152.2 | XP_006234214.1 | 12 | 1095 | |||
| XM_008761704.1 | XP_008759926.1 | 9 | 1177 | |||
| XM_008761705.1 | XP_008759927.1 | 11 | 977 | |||
| XM_008761706.1 | XP_008759928.1 | 10 | 903 | |||
| XM_008761707.1 | XP_008759929.1 | 10 | 1295 | |||
| GenBank mRNA | BC060516.1 | - | 9 | 835 | 5598062 | |
| FQ226016.1 | - | 4 | 403 | |||
| FQ230805.1 | - | 4 | 403 | |||
| FQ231958.1 | - | 4 | 403 | |||

Orc4
Genomic Map