siRNA Details
| Gene Symbol: Prpf40a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106480.1 | NP_001099950.1 | 16 | 1814 | ||
| XM_006234164.2 | XP_006234226.1 | 16 | 2139 | |||
| XM_008761709.1 | XP_008759931.1 | 16 | 2266 | |||
| XM_008761710.1 | XP_008759932.1 | 15 | 2213 | |||
| XM_008761711.1 | XP_008759933.1 | 16 | 2139 | |||
| XM_008761712.1 | XP_008759934.1 | 15 | 2086 | |||
| XM_008761713.1 | XP_008759935.1 | 15 | 1590 | |||

Prpf40a
Genomic Map