siRNA Details
| Gene Symbol: Prpf40a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106480.1 | NP_001099950.1 | 17 | 1923 | ||
| XM_006234164.2 | XP_006234226.1 | 17 | 2248 | |||
| XM_008761709.1 | XP_008759931.1 | 17 | 2375 | |||
| XM_008761710.1 | XP_008759932.1 | 16 | 2322 | |||
| XM_008761711.1 | XP_008759933.1 | 17 | 2248 | |||
| XM_008761712.1 | XP_008759934.1 | 16 | 2195 | |||
| XM_008761713.1 | XP_008759935.1 | 16 | 1699 | |||

Prpf40a
Genomic Map