siRNA Details
| Gene Symbol: Prpf40a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106480.1 | NP_001099950.1 | 20 | 2214 | ||
| XM_006234164.2 | XP_006234226.1 | 20 | 2539 | |||
| XM_008761709.1 | XP_008759931.1 | 20 | 2678 | |||
| XM_008761710.1 | XP_008759932.1 | 19 | 2625 | |||
| XM_008761711.1 | XP_008759933.1 | 20 | 2551 | |||
| XM_008761712.1 | XP_008759934.1 | 19 | 2498 | |||
| XM_008761713.1 | XP_008759935.1 | 19 | 2002 | |||

Prpf40a
Genomic Map